Product Details

SNP ID
rs141123465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8162351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGCCGGGGCAGCAGGGGGGGC[A/C]GTGGCAGCGGTAGCAGACCTGAGGA
Phenotype
MIM: 602260 MIM: 185881
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PER1 PubMed Links

Gene Details

Gene
PER1
Gene Name
period circadian clock 1
There are no transcripts associated with this gene.

Gene
VAMP2
Gene Name
vesicle associated membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014232.2 447 Silent Mutation ACG,ACT T7T NP_055047.2
XM_005256775.4 447 Silent Mutation ACG,ACT T9T XP_005256832.1

View Full Product Details