Product Details
- SNP ID
-
rs142006994
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:6758123 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTAGTCTCTGCCAACTTCACCCTC[C/T]ATGAGGCTTACTGCCTGCGGTTCCT
- Phenotype
-
MIM: 606717
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
XAF1
PubMed Links
Gene Details
- Gene
- XAF1
- Gene Name
- XIAP associated factor 1
There are no transcripts associated with this gene.
View Full Product Details