Product Details

SNP ID
rs142953484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81313772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCCTGCAAGAAAGCCAGACTGC[C/T]CCCAGGCAGGCCCTTCATGTCTTCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00482 PubMed Links

Gene Details

Gene
LINC00482
Gene Name
long intergenic non-protein coding RNA 482
There are no transcripts associated with this gene.

Gene
TMEM105
Gene Name
transmembrane protein 105
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178520.3 819 Missense Mutation GAG,GGG E90G NP_848615.1
XM_017024522.1 819 Missense Mutation GAG,GGG E68G XP_016880011.1

View Full Product Details