Product Details

SNP ID
rs143262009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18316154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTGAACTTCGTGGAGGACTCCA[A/G]GGTGGTGCTGGGAGATTCTAAGGAG
Phenotype
MIM: 601243
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMCR8 PubMed Links

Gene Details

Gene
SMCR8
Gene Name
Smith-Magenis syndrome chromosome region, candidate 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144775.2 875 Missense Mutation AAG,AGG K122R NP_658988.2
Gene
TOP3A
Gene Name
topoisomerase (DNA) III alpha
There are no transcripts associated with this gene.

View Full Product Details