Product Details

SNP ID
rs143458938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74862834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGGCTCAGTGGCCCAGGAGGC[A/G]CAGCATCTCCTGAGGATCCACCAGC
Phenotype
MIM: 103270 MIM: 138254
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FDXR PubMed Links

Gene Details

Gene
FDXR
Gene Name
ferredoxin reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258012.3 1604 Missense Mutation NP_001244941.2
NM_001258013.3 1604 Missense Mutation NP_001244942.2
NM_001258014.3 1604 Missense Mutation NP_001244943.2
NM_001258015.2 1604 Missense Mutation NP_001244944.1
NM_001258016.3 1604 Missense Mutation NP_001244945.2
NM_004110.5 1604 Missense Mutation NP_004101.3
NM_024417.4 1604 Missense Mutation NP_077728.3
XM_006721772.1 1604 Missense Mutation XP_006721835.1
XM_011524527.1 1604 Missense Mutation XP_011522829.1
XM_011524528.1 1604 Missense Mutation XP_011522830.1
XM_011524529.1 1604 Missense Mutation XP_011522831.1
XM_011524531.2 1604 Missense Mutation XP_011522833.1
XM_011524532.2 1604 Missense Mutation XP_011522834.1
XM_011524533.2 1604 Missense Mutation XP_011522835.1
Gene
GRIN2C
Gene Name
glutamate ionotropic receptor NMDA type subunit 2C
There are no transcripts associated with this gene.

View Full Product Details