Product Details

SNP ID
rs143607889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50548811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTCCCCATGGCCCTGTTCAGT[C/T]GGGTACTCCACCTGCCACTGGTGCC
Phenotype
MIM: 607264 MIM: 613939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNA1G-AS1 PubMed Links

Gene Details

Gene
CACNA1G-AS1
Gene Name
CACNA1G antisense RNA 1
There are no transcripts associated with this gene.

Gene
EPN3
Gene Name
epsin 3
There are no transcripts associated with this gene.

Gene
SPATA20
Gene Name
spermatogenesis associated 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258372.1 435 Silent Mutation GTC,GTT V105V NP_001245301.1
NM_001258373.1 435 Silent Mutation GTC,GTT V61V NP_001245302.1
NM_022827.3 435 Silent Mutation GTC,GTT V121V NP_073738.2

View Full Product Details