Product Details

SNP ID
rs144528540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49578960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCACTTCCAACACAATGCCACAG[G/T]CCAGGGAAACATCTCCATCAGCCTC
Phenotype
MIM: 604636
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100288866 PubMed Links

Gene Details

Gene
LOC100288866
Gene Name
uncharacterized LOC100288866
There are no transcripts associated with this gene.

Gene
NXPH3
Gene Name
neurexophilin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007225.2 703 Missense Mutation GGC,GTC G140V NP_009156.2

View Full Product Details