Product Details

SNP ID
rs145314397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42295722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACTGGTCACGCAGGACACAGAGA[A/T]TGAGCTGAAGAAACTGCAGCAGACT
Phenotype
MIM: 601511
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
STAT5A PubMed Links

Gene Details

Gene
STAT5A
Gene Name
signal transducer and activator of transcription 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288718.1 739 Missense Mutation AAT,ATT N160I NP_001275647.1
NM_001288719.1 739 Missense Mutation AAT,ATT N130I NP_001275648.1
NM_001288720.1 739 Missense Mutation AAT,ATT N160I NP_001275649.1
NM_003152.3 739 Missense Mutation AAT,ATT N160I NP_003143.2
XM_005257624.3 739 Missense Mutation AAT,ATT N160I XP_005257681.1

View Full Product Details