Product Details

SNP ID
rs145365732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7176988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCCCAGCGCCCTCACCCTGGGT[C/G]CTCAAGCCCTTGACCTGGGCCTCCG
Phenotype
MIM: 108360
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASGR1 PubMed Links

Gene Details

Gene
ASGR1
Gene Name
asialoglycoprotein receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197216.2 583 Missense Mutation AGC,AGG S53R NP_001184145.1
NM_001671.4 583 Missense Mutation AGC,AGG S92R NP_001662.1
XM_011523861.2 583 Missense Mutation AGC,AGG S92R XP_011522163.1

View Full Product Details