Product Details

SNP ID
rs145534754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76736270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGGAGGACTCCTGGACCGAGAC[A/C]GGGACCTGGACCGCGAACGAGATCT
Phenotype
MIM: 604914 MIM: 615262 MIM: 600813
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
JMJD6 PubMed Links

Gene Details

Gene
JMJD6
Gene Name
arginine demethylase and lysine hydroxylase
There are no transcripts associated with this gene.

Gene
METTL23
Gene Name
methyltransferase like 23
There are no transcripts associated with this gene.

Gene
MFSD11
Gene Name
major facilitator superfamily domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242532.1 808 Intron NP_001229461.1
NM_001242533.1 808 Intron NP_001229462.1
NM_001242534.1 808 Intron NP_001229463.1
NM_001242535.1 808 Intron NP_001229464.1
NM_001242536.1 808 Intron NP_001229465.1
NM_001242537.1 808 Intron NP_001229466.1
NM_024311.3 808 Intron NP_077287.1
XM_011525236.2 808 Intron XP_011523538.1
XM_011525237.2 808 Intron XP_011523539.2
XM_011525238.2 808 Intron XP_011523540.2
XM_011525239.2 808 Intron XP_011523541.2
XM_011525240.2 808 Intron XP_011523542.2
XM_011525241.2 808 Intron XP_011523543.2
XM_011525242.1 808 Intron XP_011523544.1
XM_011525244.1 808 Intron XP_011523546.1
XM_011525247.2 808 Intron XP_011523549.2
XM_017025065.1 808 Intron XP_016880554.1
XM_017025066.1 808 Intron XP_016880555.1
XM_017025067.1 808 Intron XP_016880556.1
XM_017025068.1 808 Intron XP_016880557.1
XM_017025069.1 808 Intron XP_016880558.1
XM_017025070.1 808 Intron XP_016880559.1
XM_017025071.1 808 Intron XP_016880560.1
Gene
MIR636
Gene Name
microRNA 636
There are no transcripts associated with this gene.

Gene
SRSF2
Gene Name
serine and arginine rich splicing factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195427.1 808 Missense Mutation CGG,CTG R186L NP_001182356.1
NM_003016.4 808 Missense Mutation CGG,CTG R186L NP_003007.2
XM_017024942.1 808 Missense Mutation CGG,CTG R186L XP_016880431.1

View Full Product Details