Product Details

SNP ID
rs146159563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41149148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGGAAGTGTTCATTTTTCCAG[C/T]GACTGGCTGGCTACAGTGCATTTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP4-5 PubMed Links

Gene Details

Gene
KRTAP4-5
Gene Name
keratin associated protein 4-5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033188.3 655 UTR 3 NP_149445.3
Gene
KRTAP4-6
Gene Name
keratin associated protein 4-6
There are no transcripts associated with this gene.

View Full Product Details