Product Details

SNP ID
rs146417493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28673355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGTCCGTTTTTTTATCCTTCAC[G/T]CTTTTCTTTCCCTAGTTATCTTCAG
Phenotype
MIM: 601333
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SUPT6H PubMed Links

Gene Details

Gene
SUPT6H
Gene Name
SPT6 homolog, histone chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320755.1 Intron NP_001307684.1
NM_003170.4 Intron NP_003161.2
XM_017024978.1 Intron XP_016880467.1
XM_017024979.1 Intron XP_016880468.1
XM_017024980.1 Intron XP_016880469.1

View Full Product Details