Product Details

SNP ID
rs147453724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64858828 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAATCGTTAGTATTCCAGTCACA[A/G]TTAACGCCAAGATGAGTTTTTTGGT
Phenotype
MIM: 616557
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC37A3 PubMed Links

Gene Details

Gene
LRRC37A3
Gene Name
leucine rich repeat containing 37 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303255.1 5426 Missense Mutation NP_001290184.1
NM_199340.3 5426 Missense Mutation NP_955372.2
XM_011524776.1 5426 Intron XP_011523078.1
XM_011524777.1 5426 Intron XP_011523079.1
XM_011524778.2 5426 Intron XP_011523080.2
XM_017024593.1 5426 Intron XP_016880082.1
XM_017024594.1 5426 Intron XP_016880083.1
XM_017024595.1 5426 Intron XP_016880084.1
XM_017024596.1 5426 Intron XP_016880085.1
XM_017024597.1 5426 Intron XP_016880086.1
XM_017024598.1 5426 Intron XP_016880087.1
XM_017024599.1 5426 Intron XP_016880088.1
XM_017024600.1 5426 Intron XP_016880089.1
XM_017024601.1 5426 Missense Mutation ACT,ATT T851I XP_016880090.1
XM_017024602.1 5426 Missense Mutation ACT,ATT T776I XP_016880091.1
XM_017024603.1 5426 Intron XP_016880092.1
XM_017024604.1 5426 Intron XP_016880093.1
XM_017024605.1 5426 Intron XP_016880094.1
XM_017024606.1 5426 Intron XP_016880095.1
XM_017024607.1 5426 Intron XP_016880096.1
XM_017024608.1 5426 Intron XP_016880097.1

View Full Product Details