Product Details
- SNP ID
-
rs147465833
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:7075093 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTGGTCTGACCCAGGCCAGCCTC[A/G]CAGACCCAGTGGTAGGGCCTCTGGC
- Phenotype
-
MIM: 605999
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLEC10A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs15623,rs78425351] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLEC10A
- Gene Name
- C-type lectin domain family 10 member A
View Full Product Details