Product Details

SNP ID
rs147846226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16940339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGCACTCACCCTGGGAAGACTT[A/G]GCCGGACTTTGACGGGGCCTTGAGC
Phenotype
MIM: 604907
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF13B PubMed Links

Gene Details

Gene
TNFRSF13B
Gene Name
TNF receptor superfamily member 13B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012452.2 631 Silent Mutation GCC,GCT A206A NP_036584.1

View Full Product Details