Product Details

SNP ID
rs149053045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39407585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGATAAGATTGTGTTACTTGTCA[C/T]AGACAAGGACTGGTCTGAAGAGATG
Phenotype
MIM: 609086 MIM: 604311
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXL20 PubMed Links

Gene Details

Gene
FBXL20
Gene Name
F-box and leucine rich repeat protein 20
There are no transcripts associated with this gene.

Gene
MED1
Gene Name
mediator complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004774.3 4781 Missense Mutation ATG,GTG M1546V NP_004765.2
XM_005257465.3 4781 Missense Mutation ATG,GTG M1374V XP_005257522.1
XM_006721957.1 4781 Intron XP_006722020.1
XM_017024779.1 4781 Missense Mutation ATG,GTG M1327V XP_016880268.1

View Full Product Details