Product Details

SNP ID
rs149253093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34271250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGACCACCAGTAGCCACTGTCCC[C/T]GGGAAGCTGTAATGTATGTGGACGA
Phenotype
MIM: 158106
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL7 PubMed Links

Gene Details

Gene
CCL7
Gene Name
C-C motif chemokine ligand 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006273.3 256 Missense Mutation CGG,TGG R61W NP_006264.2

View Full Product Details