Product Details

SNP ID
rs149554562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75898926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCAGGGGCTGCCGGTGGGGGAAG[C/T]GCTTCTGCTTGGGGTGGTAAGGGGG
Phenotype
MIM: 611844
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL38 PubMed Links

Gene Details

Gene
MRPL38
Gene Name
mitochondrial ribosomal protein L38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032478.3 1292 Missense Mutation CAC,CGC H356R NP_115867.2
Gene
TRIM65
Gene Name
tripartite motif containing 65
There are no transcripts associated with this gene.

View Full Product Details