Product Details

SNP ID
rs149746467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7075139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCAGACGTCGTCATTCCACCTG[A/C]CGTCTGGATGGAAGTGAGCACAGTC
Phenotype
MIM: 605999
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLEC10A PubMed Links

Gene Details

Gene
CLEC10A
Gene Name
C-type lectin domain family 10 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005256412.1 1204 Missense Mutation XP_005256469.1
XM_011523613.1 1204 Missense Mutation XP_011521915.1
XM_011523614.2 1204 Missense Mutation XP_011521916.1
XM_011523615.1 1204 Missense Mutation XP_011521917.1

View Full Product Details