Product Details

SNP ID
rs150188248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21198690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGCCCACGGTGATCCAGCGGGC[C/T]GTCTCGTCGCCAATGCGAGTGGGCT
Phenotype
MIM: 616160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHRS7B PubMed Links

Gene Details

Gene
DHRS7B
Gene Name
dehydrogenase/reductase 7B
There are no transcripts associated with this gene.

Gene
TMEM11
Gene Name
transmembrane protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003876.2 656 Silent Mutation ACA,ACG T71T NP_003867.1

View Full Product Details