Product Details

SNP ID
rs150816372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38937237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCTTATGGAAGTTAGCCTGAG[C/T]ATGTACAAGATGGAACAAATTCAAA
Phenotype
MIM: 609498
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO47 PubMed Links

Gene Details

Gene
FBXO47
Gene Name
F-box protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008777.2 1399 Missense Mutation ACT,GCT T433A NP_001008777.2
XM_011524865.2 1399 Missense Mutation ACT,GCT T407A XP_011523167.1
XM_011524866.2 1399 Missense Mutation ACT,GCT T376A XP_011523168.1
XM_011524867.2 1399 Intron XP_011523169.1
Gene
LINC00672
Gene Name
long intergenic non-protein coding RNA 672
There are no transcripts associated with this gene.

View Full Product Details