Product Details

SNP ID
rs150890641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39630043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGCCTACACACCACCTTCGCT[A/G]AAAGGTACTATACCCCCACCGACCT
Phenotype
MIM: 604399 MIM: 607048
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R1B PubMed Links

Gene Details

Gene
PPP1R1B
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242464.1 1057 Silent Mutation CTA,CTG L43L NP_001229393.1
NM_032192.3 1057 Silent Mutation CTA,CTG L79L NP_115568.2
NM_181505.3 1057 Silent Mutation CTA,CTG L43L NP_852606.1
XM_017025216.1 1057 Silent Mutation CTA,CTG L79L XP_016880705.1
XM_017025217.1 1057 Silent Mutation CTA,CTG L43L XP_016880706.1
Gene
STARD3
Gene Name
StAR related lipid transfer domain containing 3
There are no transcripts associated with this gene.

View Full Product Details