Product Details

SNP ID
rs150946853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64967739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGCAAAAGAAAAAAAATGAGCT[A/G]ATGCACTCGAGAGAGTAAGGAAGGC
Phenotype
MIM: 616557
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMZ2P1 PubMed Links

Gene Details

Gene
AMZ2P1
Gene Name
archaelysin family metallopeptidase 2 pseudogene 1
There are no transcripts associated with this gene.

Gene
LRRC37A3
Gene Name
leucine rich repeat containing 37 member A3
There are no transcripts associated with this gene.

View Full Product Details