Product Details

SNP ID
rs143898455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50974300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAGACCAAGAATCCAGACTGTT[A/C]TAAATAACCAAAAAGTAATAATAGA
Phenotype
MIM: 608079
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ELAC1 PubMed Links

Gene Details

Gene
ELAC1
Gene Name
elaC ribonuclease Z 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018696.2 Intron NP_061166.1

View Full Product Details