Product Details

SNP ID
rs145679761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12955588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAGTAGGAGAATCAAATGATAA[A/G]CTGCGAGGACAGAGCCACTGGGTGA
Phenotype
MIM: 609263
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEH1L PubMed Links

Gene Details

Gene
SEH1L
Gene Name
SEH1 like nucleoporin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013437.1 596 Silent Mutation AAA,AAG K96K NP_001013455.1
NM_031216.3 596 Silent Mutation AAA,AAG K96K NP_112493.2
XM_005258152.4 596 Intron XP_005258209.1
XM_011525742.2 596 Silent Mutation AAA,AAG K96K XP_011524044.1
XM_011525743.2 596 Intron XP_011524045.1
XM_011525744.1 596 Silent Mutation AAA,AAG K96K XP_011524046.1
XM_011525745.2 596 Intron XP_011524047.1
XM_011525746.2 596 UTR 5 XP_011524048.1
XM_017026025.1 596 Intron XP_016881514.1
XM_017026026.1 596 Intron XP_016881515.1
XM_017026027.1 596 UTR 5 XP_016881516.1

View Full Product Details