Product Details

SNP ID
rs146107834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50886697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTCTGTTTAATGCTTTTGATGT[C/T]TACCTAGAGAAAACCAAATCTGGCT
Phenotype
MIM: 154270
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ME2 PubMed Links

Gene Details

Gene
ME2
Gene Name
malic enzyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168335.1 Intron NP_001161807.1
NM_002396.4 Intron NP_002387.1

View Full Product Details