Product Details

SNP ID
rs146610138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:1268921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATGTTATGACTTCTTCTTGACC[A/G]TTTAAACATATGCACATGCTCTGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00470 PubMed Links

Gene Details

Gene
LINC00470
Gene Name
long intergenic non-protein coding RNA 470
There are no transcripts associated with this gene.

View Full Product Details