Product Details

SNP ID
rs148461454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54544124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGGGTCAACTATGAGCATGTAA[A/G]TGCATCATAGGAAGGCAATATGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYNAP PubMed Links

Gene Details

Gene
DYNAP
Gene Name
dynactin associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307955.1 Intron NP_001294884.1
NM_173629.2 Intron NP_775900.1
XM_011525923.2 Intron XP_011524225.1
XM_011525924.2 Intron XP_011524226.1
XM_011525925.2 Intron XP_011524227.1
XM_017025709.1 Intron XP_016881198.1

View Full Product Details