Product Details

SNP ID
rs148785644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:80160250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCCACCTCAATGCCGTTCACCT[A/C]CAGGACCTCGTCATTCACAGCCAGC
Phenotype
MIM: 608976
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PARD6G PubMed Links

Gene Details

Gene
PARD6G
Gene Name
par-6 family cell polarity regulator gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032510.3 818 Nonsense Mutation GAG,TAG E218* NP_115899.1
Gene
PARD6G-AS1
Gene Name
PARD6G antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details