Product Details

SNP ID
rs138123319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:677921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGTCCAACCTCACCCACCCGG[A/G]GAACAAGATCAACCTCCTCGGCTTC
Phenotype
MIM: 605343
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FSTL3 PubMed Links

Gene Details

Gene
FSTL3
Gene Name
follistatin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005860.2 268 Missense Mutation GAG,GGG E78G NP_005851.1
Gene
PRSS57
Gene Name
protease, serine 57
There are no transcripts associated with this gene.

View Full Product Details