Product Details

SNP ID
rs138352262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18931235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCACAGGACTGGGCCTCCCAA[C/T]ACTTTATCATCTTAGGTAGATATTG
Phenotype
MIM: 604800
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX49 PubMed Links

Gene Details

Gene
DDX49
Gene Name
DEAD-box helicase 49
There are no transcripts associated with this gene.

Gene
HOMER3
Gene Name
homer scaffolding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145721.1 Intron NP_001139193.1
NM_001145722.1 Intron NP_001139194.1
NM_001145724.1 Intron NP_001139196.1
NM_004838.3 Intron NP_004829.3
XM_006722943.1 Intron XP_006723006.1
XM_006722944.1 Intron XP_006723007.1
Gene
LOC102724360
Gene Name
uncharacterized LOC102724360
There are no transcripts associated with this gene.

View Full Product Details