Product Details

SNP ID
rs138503106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15452033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGACCTGCCCCAGGGCTCAGA[C/T]GGCAATCTCACCAGGTTTTTCAGAC
Phenotype
MIM: 616561
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR1470 PubMed Links

Gene Details

Gene
MIR1470
Gene Name
microRNA 1470
There are no transcripts associated with this gene.

Gene
RASAL3
Gene Name
RAS protein activator like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022904.1 Intron NP_075055.1
XM_011528185.1 Intron XP_011526487.1
XM_011528186.1 Intron XP_011526488.1
XM_011528187.1 Intron XP_011526489.1
Gene
WIZ
Gene Name
widely interspaced zinc finger motifs
There are no transcripts associated with this gene.

View Full Product Details