Product Details

SNP ID
rs140442555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44781044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACCTTCTGGAGGGAAAGTTGC[A/G]GAGCCCGGTGAGTAAGCCCTTGTCC
Phenotype
MIM: 608453
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBLC PubMed Links

Gene Details

Gene
CBLC
Gene Name
Cbl proto-oncogene C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130852.1 570 Missense Mutation AGA,GGA R165G NP_001124324.1
NM_012116.3 570 Missense Mutation AGA,GGA R165G NP_036248.3
XM_005258696.3 570 Missense Mutation AGA,GGA R165G XP_005258753.1
XM_011526688.2 570 Missense Mutation AGA,GGA R165G XP_011524990.1
XM_011526689.2 570 Missense Mutation AGA,GGA R165G XP_011524991.1
XM_011526690.2 570 Missense Mutation AGA,GGA R165G XP_011524992.1

View Full Product Details