Product Details

SNP ID
rs140474922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52373486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGCCCCTTTGAGATTCATCATT[A/G]AGTGGTACCAGGGCTTAAGTATGCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF610 PubMed Links

Gene Details

Gene
ZNF610
Gene Name
zinc finger protein 610
There are no transcripts associated with this gene.

Gene
ZNF880
Gene Name
zinc finger protein 880
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145434.1 Intron NP_001138906.1
XM_017026812.1 Intron XP_016882301.1

View Full Product Details