Product Details

SNP ID
rs141018705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49635824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTAGGCCACGTGGTGGGAGGCG[C/T]CGAAGGCAGAGGCTTCTGATCGGGG
Phenotype
MIM: 616633 MIM: 165090
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRR12 PubMed Links

Gene Details

Gene
PRR12
Gene Name
proline rich 12
There are no transcripts associated with this gene.

Gene
RRAS
Gene Name
related RAS viral (r-ras) oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006270.4 527 Missense Mutation GAC,GGC D161G NP_006261.1
Gene
SCAF1
Gene Name
SR-related CTD associated factor 1
There are no transcripts associated with this gene.

View Full Product Details