Product Details

SNP ID
rs141077776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42588956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAATACTGTGAGGGTGGATTAGA[A/G]GCCGCATGGCAGGAGAGGTTGAGAT
Phenotype
MIM: 615747
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEACAM8 PubMed Links

Gene Details

Gene
CEACAM8
Gene Name
carcinoembryonic antigen related cell adhesion molecule 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001816.3 614 Silent Mutation GCC,GCT A262A NP_001807.2
XM_011526340.2 614 Silent Mutation GCC,GCT A262A XP_011524642.1
XM_011526341.1 614 Intron XP_011524643.1
XM_011526342.1 614 Intron XP_011524644.1
XM_017026194.1 614 Silent Mutation GCC,GCT A262A XP_016881683.1
XM_017026195.1 614 Silent Mutation GCC,GCT A262A XP_016881684.1
XM_017026196.1 614 Silent Mutation GCC,GCT A262A XP_016881685.1
XM_017026197.1 614 Silent Mutation GCC,GCT A142A XP_016881686.1
XM_017026198.1 614 Intron XP_016881687.1
Gene
LIPE-AS1
Gene Name
LIPE antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details