Product Details

SNP ID
rs141306136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16501221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTGGGGGTGCACTCGAACTCGC[A/G]TCCCAGAACACAGACAAAACTTCCC
Phenotype
MIM: 611414
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf44 PubMed Links

Gene Details

Gene
C19orf44
Gene Name
chromosome 19 open reading frame 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288834.1 585 Silent Mutation GCA,GCG A143A NP_001275763.1
NM_032207.3 585 Silent Mutation GCA,GCG A143A NP_115583.1
XM_006722920.3 585 Silent Mutation GCA,GCG A143A XP_006722983.1
XM_011528354.2 585 Silent Mutation GCA,GCG A143A XP_011526656.1
XM_017027350.1 585 Silent Mutation GCA,GCG A143A XP_016882839.1
Gene
CALR3
Gene Name
calreticulin 3
There are no transcripts associated with this gene.

View Full Product Details