Product Details

SNP ID
rs141519703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11575205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCTTTGAGGGGTCCATGAAATT[C/T]CCAGCCCCACTCAGCACGTAGCCCA
Phenotype
MIM: 171640 MIM: 612248
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACP5 PubMed Links

Gene Details

Gene
ACP5
Gene Name
acid phosphatase 5, tartrate resistant
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111034.2 1186 Silent Mutation GGA,GGG G261G NP_001104504.1
NM_001111035.2 1186 Silent Mutation GGA,GGG G261G NP_001104505.1
NM_001111036.2 1186 Silent Mutation GGA,GGG G261G NP_001104506.1
NM_001322023.1 1186 Silent Mutation GGA,GGG G261G NP_001308952.1
NM_001611.4 1186 Silent Mutation GGA,GGG G261G NP_001602.1
XM_005259938.1 1186 Silent Mutation GGA,GGG G261G XP_005259995.1
XM_011528069.2 1186 Silent Mutation GGA,GGG G261G XP_011526371.1
Gene
ZNF627
Gene Name
zinc finger protein 627
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290083.1 1186 Intron NP_001277012.1
NM_001290084.1 1186 Intron NP_001277013.1
NM_001290085.1 1186 Intron NP_001277014.1
NM_145295.3 1186 Intron NP_660338.1
XM_011527780.2 1186 Intron XP_011526082.1

View Full Product Details