Product Details

SNP ID
rs142215917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50512367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTTAGCTCTGCTCACCTGGTAT[C/G]AGCCACGAGCACCCCAATCACCCCA
Phenotype
MIM: 614545 MIM: 615324
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASPDH PubMed Links

Gene Details

Gene
ASPDH
Gene Name
aspartate dehydrogenase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024656.2 708 Missense Mutation CAT,GAT H111D NP_001019827.2
NM_001114598.1 708 Missense Mutation CAT,GAT H216D NP_001108070.1
Gene
EMC10
Gene Name
ER membrane protein complex subunit 10
There are no transcripts associated with this gene.

Gene
JOSD2
Gene Name
Josephin domain containing 2
There are no transcripts associated with this gene.

Gene
LRRC4B
Gene Name
leucine rich repeat containing 4B
There are no transcripts associated with this gene.

View Full Product Details