Product Details
- SNP ID
-
rs142490331
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:51414864 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGATCCACTCTGGGCCCCATGGAC[A/G]TTCCAGGCCTCACAGCGGAGCCTGA
- Phenotype
-
MIM: 606091
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100129083
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3810099] are located under a probe and SNP(s) [rs1833785] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100129083
- Gene Name
- uncharacterized LOC100129083
There are no transcripts associated with this gene.
- Gene
- SIGLEC10
- Gene Name
- sialic acid binding Ig like lectin 10
View Full Product Details