Product Details

SNP ID
rs142584501
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35225176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCCGGAGAGCTGGCTCTCCCA[A/C]GTCAGGGGGGTGTCGTTGGCACGCC
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
FAM187B PubMed Links

Gene Details

Gene
FAM187B
Gene Name
family with sequence similarity 187 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152481.1 804 Silent Mutation ACG,ACT T253T NP_689694.1

View Full Product Details