Product Details

SNP ID
rs143148835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49819238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTGCGCTCCCGAGTCCTACGTA[C/G]AATGTCACGCTCCCACCAGCAGCGC
Phenotype
MIM: 610622 MIM: 610197
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FUZ PubMed Links

Gene Details

Gene
FUZ
Gene Name
fuzzy planar cell polarity protein
There are no transcripts associated with this gene.

Gene
MED25
Gene Name
mediator complex subunit 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030973.3 310 Missense Mutation CAA,GAA Q83E NP_112235.2
XM_011527353.1 310 Missense Mutation CAA,GAA Q83E XP_011525655.1

View Full Product Details