Product Details

SNP ID
rs143847450
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9566903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAACATTTGGTGGCAGGCTGAA[A/G]GCTTTTCTGCACTGATTCAACACAG
Phenotype
MIM: 194628
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF121 PubMed Links

Gene Details

Gene
ZNF121
Gene Name
zinc finger protein 121
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008727.3 442 Silent Mutation GCC,GCT A70A NP_001008727.1
NM_001308269.1 442 Silent Mutation GCC,GCT A70A NP_001295198.1
XM_017027239.1 442 Silent Mutation GCC,GCT A70A XP_016882728.1

View Full Product Details