Product Details

SNP ID
rs143937870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51413805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGGTTTCTGTCTGAGTCCGTCT[C/T]TTCGGTAGAATCTTCATGCTGAGGA
Phenotype
MIM: 606091
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100129083 PubMed Links

Gene Details

Gene
LOC100129083
Gene Name
uncharacterized LOC100129083
There are no transcripts associated with this gene.

Gene
SIGLEC10
Gene Name
sialic acid binding Ig like lectin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171156.1 1491 Silent Mutation AAA,AAG K518K NP_001164627.1
NM_001171157.1 1491 Intron NP_001164628.1
NM_001171158.1 1491 Silent Mutation AAA,AAG K433K NP_001164629.1
NM_001171159.1 1491 Silent Mutation AAA,AAG K423K NP_001164630.1
NM_001171161.1 1491 Silent Mutation AAA,AAG K333K NP_001164632.1
NM_001322105.1 1491 Silent Mutation AAA,AAG K391K NP_001309034.1
NM_033130.4 1491 Silent Mutation AAA,AAG K576K NP_149121.2
XM_005259366.4 1491 Silent Mutation AAA,AAG K486K XP_005259423.1
XM_005259367.3 1491 Silent Mutation AAA,AAG K428K XP_005259424.1
XM_011527439.1 1491 Silent Mutation AAA,AAG K528K XP_011525741.1

View Full Product Details