Product Details

SNP ID
rs144148015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3179152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGGCGCCCGCCCAGTGGTTCCT[A/G]CGGGAGGGCCTGCTCTTCACCGCCC
Phenotype
MIM: 609156 MIM: 603751
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCLN PubMed Links

Gene Details

Gene
NCLN
Gene Name
nicalin
There are no transcripts associated with this gene.

Gene
S1PR4
Gene Name
sphingosine-1-phosphate receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003775.3 415 Silent Mutation CTA,CTG L120L NP_003766.1

View Full Product Details