Product Details
- SNP ID
-
rs144306949
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37413055 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCTCAAATGAAGGGTAAGGGATG[A/C]AATTTGAGAGAAGGCTTTACCACAT
- Phenotype
-
MIM: 613904
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF569
PubMed Links
Gene Details
- Gene
- ZNF569
- Gene Name
- zinc finger protein 569
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152484.2 |
1744 |
Missense Mutation |
GCA,TCA |
A535S |
NP_689697.2 |
XM_006723046.2 |
1744 |
Missense Mutation |
GCA,TCA |
A559S |
XP_006723109.1 |
XM_006723047.3 |
1744 |
Missense Mutation |
GCA,TCA |
A559S |
XP_006723110.1 |
XM_006723048.3 |
1744 |
Missense Mutation |
GCA,TCA |
A559S |
XP_006723111.1 |
XM_011526538.2 |
1744 |
Missense Mutation |
GCA,TCA |
A559S |
XP_011524840.1 |
XM_011526539.2 |
1744 |
Missense Mutation |
GCA,TCA |
A535S |
XP_011524841.1 |
XM_017026376.1 |
1744 |
Missense Mutation |
GCA,TCA |
A535S |
XP_016881865.1 |
XM_017026377.1 |
1744 |
Missense Mutation |
GCA,TCA |
A535S |
XP_016881866.1 |
XM_017026378.1 |
1744 |
Missense Mutation |
GCA,TCA |
A499S |
XP_016881867.1 |
XM_017026379.1 |
1744 |
Missense Mutation |
GCA,TCA |
A376S |
XP_016881868.1 |
XM_017026380.1 |
1744 |
Missense Mutation |
GCA,TCA |
A376S |
XP_016881869.1 |
XM_017026381.1 |
1744 |
Missense Mutation |
GCA,TCA |
A376S |
XP_016881870.1 |
XM_017026382.1 |
1744 |
Missense Mutation |
GCA,TCA |
A376S |
XP_016881871.1 |
View Full Product Details