Product Details

SNP ID
rs144386323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15227462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGTCCAGCAGGTCTTGCAAGAAG[A/G]CCCACATGTACTGGTGCATCTCCTG
Phenotype
MIM: 608749
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRD4 PubMed Links

Gene Details

Gene
BRD4
Gene Name
bromodomain containing 4
There are no transcripts associated with this gene.

Gene
EPHX3
Gene Name
epoxide hydrolase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142886.1 1267 Missense Mutation GCC,GTC A353V NP_001136358.1
NM_024794.2 1267 Missense Mutation GCC,GTC A353V NP_079070.1

View Full Product Details