Product Details

SNP ID
rs144556033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2477502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCTACAGAACCCTCACACGGA[C/T]GCCTGGAAGAGCCACGGCTTGGTGG
Phenotype
MIM: 604948
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GADD45B PubMed Links

Gene Details

Gene
GADD45B
Gene Name
growth arrest and DNA damage inducible beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015675.3 618 Silent Mutation GAC,GAT D128D NP_056490.2
XM_017026822.1 618 UTR 3 XP_016882311.1

View Full Product Details