Product Details

SNP ID
rs144687950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52801073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGAGATCTACGATGGCATGCAA[A/G]GTATCGCTTCTGATTAAATACCTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF28 PubMed Links

Gene Details

Gene
ZNF28
Gene Name
zinc finger protein 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006969.3 959 Intron NP_008900.3
XM_006723354.3 959 Intron XP_006723417.1
XM_011527262.2 959 Missense Mutation CTT,TTT L258F XP_011525564.1

View Full Product Details