Product Details

SNP ID
rs144768117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4660002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTCCCTTTCAAATGCGGCTTTA[C/G]ACTGAAAAAGAATGAGTGGAAACTT
Phenotype
MIM: 606746 MIM: 615869
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYDGF PubMed Links

Gene Details

Gene
MYDGF
Gene Name
myeloid derived growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019107.3 440 Missense Mutation TCT,TGT S124C NP_061980.1
XM_017026987.1 440 Intron XP_016882476.1
Gene
TNFAIP8L1
Gene Name
TNF alpha induced protein 8 like 1
There are no transcripts associated with this gene.

View Full Product Details